1,1,1,2,3,3-Hexafluoro-2-(heptafluoropropoxy)-3-[(trifluorovinyl)oxy]propane ≥97%

Supplier: Matrix Scientific

Synonyms: Perfluoro(5-methyl-3,6-dioxanon-1-ene)

009116-1G 009116-5G
101796-396EA 32.31 USD
101796-396 101796-398
1,1,1,2,3,3-Hexafluoro-2-(heptafluoropropoxy)-3-[(trifluorovinyl)oxy]propane ≥97%
1,1,1,2,3,3-Hexafluoro-2-(heptafluoropropoxy)-3-[(trifluorovinyl)oxy]propane

Hazard: Irritant

Formula: F₂CCFOCF₂CF(CF₃)O(CF₂)₂CF₃
MW: 432.06 g/mol
Boiling Pt: 103°C
Density: 1.8
MDL Number: MFCD00191479
CAS Number: 1644-11-7

Order Now

Specification Test Results

Density 1.8

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR