3-Chloro-2-methylbenzylamine

Supplier: Matrix Scientific

002171-1G 002171-5G
101783-378EA 32.31 USD
101783-378 101783-380
3-Chloro-2-methylbenzylamine
3-Chloro-2-methylbenzylamine

Hazard: Irritant

Formula: C₈H₁₀ClN
Density: 1.14
MDL Number: MFCD00053644
CAS Number: 226565-61-3

Order Now

Specification Test Results

Density 1.14

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR