2,2,2-Trifluoroethyl methanesulfonate ≥97%
Supplier: Matrix Scientific
003705-25G
003705-100G
101786-100EA
112.31
USD
101786-100
101786-102
2,2,2-Trifluoroethyl methanesulfonate ≥97%
2,2,2-Trifluoroethyl methanesulfonate
Hazard: Irritant
Formula:
C₃H₅F₃O₃S Density: 1.52 |
MDL Number:
MFCD00025058 CAS Number: 25236-64-0 |
Specification Test Results
Density | 1.52 |
ND20 | 1.367 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...