1H,1H,9H-Hexadecafluorononyl acrylate ≥97%

Supplier: Matrix Scientific

Synonyms: 1H,1H,9H-Perfluorononyl acrylate

002833-5G 002833-25G
101784-746EA 223.08 USD
101784-746 101784-748
1H,1H,9H-Hexadecafluorononyl acrylate ≥97%
1H,1H,9H-Hexadecafluorononyl acrylate

Hazard: Irritant

Formula: C₁₂H₆F₁₆O₂
MW: 486.15 g/mol
Density: 1.646
MDL Number: MFCD00078281
CAS Number: 4180-26-1

Order Now

Specification Test Results

Density 1.646
ND20 1.349

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR