2'-Fluoro-5'-(trifluoromethyl)acetophenone ≥98%

Supplier: Matrix Scientific

001496-5G 001496-25G
101781-818EA 81.54 USD
101781-818 101781-820
2'-Fluoro-5'-(trifluoromethyl)acetophenone ≥98%
2'-Fluoro-5'-(trifluoromethyl)acetophenone

Hazard: Irritant

Formula: C₉H₆F₄O
MW: 206.14 g/mol
MDL Number: MFCD00061248
CAS Number: 202664-53-7

Order Now

Specification Test Results

ND20 1.447

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR