2'-Fluoro-3'-(trifluoromethyl)acetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2-Fluoro-3-(trifluoromethyl)phenyl)ethanone

001437-1G 001437-5G
101781-644EA 23.08 USD
101781-644 101781-646
2'-Fluoro-3'-(trifluoromethyl)acetophenone ≥98%
2'-Fluoro-3'-(trifluoromethyl)acetophenone

Hazard: Irritant

Formula: C₉H₆F₄O
MW: 206.14 g/mol
Density: 1.02
MDL Number: MFCD00061145
CAS Number: 207853-63-2

Order Now

Specification Test Results

Density 1.02
ND20 1.453

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR