2-(Chloromethyl)-4-(trifluoromethyl)pyridine ≥95%

Supplier: Matrix Scientific

070777-500MG 070777-1G
101946-286EA 889.23 USD
101946-286 101946-288
2-(Chloromethyl)-4-(trifluoromethyl)pyridine ≥95%
2-(Chloromethyl)-4-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₇H₅ClF₃N
Boiling Pt: 199 °C
MDL Number: MFCD10697599
CAS Number: 215867-87-1

Order Now

Specification Test Results

Boiling point 199°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR