1-Bromo-4-(bromomethyl)-2-chlorobenzene ≥95%
Supplier: Matrix Scientific
066633-1G
066633-5G
101932-032EA
43.08
USD
101932-032
101932-034
1-Bromo-4-(bromomethyl)-2-chlorobenzene ≥95%
1-Bromo-4-(bromomethyl)-2-chlorobenzene
Hazard: Irritant
Formula:
C₇H₅Br₂Cl Boiling Pt: 90°C/0.4mm |
MDL Number:
MFCD08059101 CAS Number: 68120-44-5 |
Specification Test Results
Boiling point | 90°C/0.4mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...