2-Chloro-5-fluorobenzotrifluoride ≥95%
Supplier: Matrix Scientific
064050-1G
064050-5G
101926-890EA
15.38
USD
101926-890
101926-892
2-Chloro-5-fluorobenzotrifluoride ≥95%
2-Chloro-5-fluorobenzotrifluoride
Hazard: Irritant
Formula:
C₇H₃ClF₄ MW: 198.5500 g/mol Boiling Pt: 76°C/80mm |
MDL Number:
MFCD07782058 CAS Number: 89634-75-3 |
Specification Test Results
Boiling point | 76°C/80mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...