Vinylphenylacetonitrile

Supplier: Matrix Scientific

057213-2G 057213-50G
101912-772EA 143.08 USD
101912-772 101912-776
Vinylphenylacetonitrile
Vinylphenylacetonitrile

Mixture of 30% para, 70% meta


Hazard: Irritant

Formula: C₁₀H₉N
Boiling Pt: 108 °C (1 mmHg)
MDL Number: MFCD17168382
CAS Number: 58879-50-8

Order Now

Specification Test Results

Boiling point 108°C/1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR