2,5-Dimethylpyrrole

Supplier: Matrix Scientific

Synonyms: 2,5-Dimethyl-1H-pyrrole

057049-5G 057049-25G
101912-178EA 216.92 USD
101912-178 101912-180
2,5-Dimethylpyrrole
2,5-Dimethylpyrrole

Hazard: Irritant

Formula: C₆H₉N
MW: 95.1424 g/mol
Boiling Pt: 51-53°C/8mm
MDL Number: MFCD00005223
CAS Number: 625-84-3

Order Now

Specification Test Results

Boiling point 51-53°C/8mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR