3,4-Dimethylfurazan

Supplier: Matrix Scientific

057035-5G 057035-25G
101912-122EA 238.46 USD
101912-122 101912-124
3,4-Dimethylfurazan
3,4-Dimethylfurazan

Hazard: Irritant

Formula: C₄H₆N₂O
Boiling Pt: 157 °C
MDL Number: MFCD00020815
CAS Number: 4975-21-7

Order Now

Specification Test Results

Boiling point 157°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR