α,2,3-Trichlorotoluene

Supplier: Matrix Scientific

Synonyms: 1,2-Dichloro-3-(chloromethyl)benzene, alpha,2,3-Trichlorotoluene, 1-(Chloromethyl)-2,3-dichlorobenzene, 2,3-Dichlorobenzyl chloride

057005-5G 057005-25G
101912-014EA 210.77 USD
101912-014 101912-016
α,2,3-Trichlorotoluene
α,2,3-Trichlorotoluene

Hazard: Irritant/Lachrymator

Formula: C₇H₅Cl₃
MW: 195.47 g/mol
Boiling Pt: 255 °C
MDL Number: MFCD00035817
CAS Number: 3290-01-5

Order Now

Specification Test Results

Boiling point 255


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR