2-Chlorobenzyl thiocyanate ≥98%

Supplier: Matrix Scientific

056947-10G 056947-25G 056947-100G
101911-826EA 176.92 USD
101911-826 101911-828 101911-830
2-Chlorobenzyl thiocyanate ≥98%
2-Chlorobenzyl thiocyanate

Hazard: Irritant-Harmful

Formula: C₈H₆ClNS
MW: 183.66 g/mol
Boiling Pt: 113 °C (1 mmHg)
MDL Number: MFCD09025671
CAS Number: 2082-66-8

Order Now

Specification Test Results

Boiling point 113°C/1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR