4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone ≥95%
Supplier: Matrix Scientific
048519-500MG
048519-1G
101884-186EA
556.92
USD
101884-186
101884-188
4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone ≥95%
4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone
Hazard: Irritant
Formula:
C₈H₁₀Br₂N₂O MW: 310 g/mol Boiling Pt: 180°C (3 mmHg) |
MDL Number:
MFCD12079183 CAS Number: 107784-73-6 |
Specification Test Results
Boiling point | 180@3mmHg°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...