2-(2-Chloro-6-methoxy-4-pyridinyl)-2-propanol ≥90%

Supplier: Matrix Scientific

048091-500MG 048091-1G
101882-760EA 556.92 USD
101882-760 101882-762
2-(2-Chloro-6-methoxy-4-pyridinyl)-2-propanol ≥90%
2-(2-Chloro-6-methoxy-4-pyridinyl)-2-propanol

Hazard: Irritant

Formula: C₉H₁₂ClNO₂
Boiling Pt: 109 °C (0.8 mmHg)
MDL Number: MFCD11841027
CAS Number: 95037-52-8

Order Now

Specification Test Results

Boiling point 109°C/0.8mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR