Ethyl-1-benzofuran-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl benzofuran-2-carboxylate

047965-500MG 047965-1G
101882-194EA 20 USD
101882-194 101882-196
Ethyl-1-benzofuran-2-carboxylate ≥95%
Ethyl-1-benzofuran-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
Boiling Pt: 105°C/0.1mm
MDL Number: MFCD03094950
CAS Number: 3199-61-9

Order Now

Specification Test Results

Boiling point 105°C/0.1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR