1-Isopentyltetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

047919-500MG 047919-1G 047919-5G
101881-998EA 276.92 USD
101881-998 101882-000 101882-002
1-Isopentyltetrahydro-4(1H)-pyridinone ≥95%
1-Isopentyltetrahydro-4(1H)-pyridinone

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₉NO
Boiling Pt: 60 °C (0.3 mmHg)
MDL Number: MFCD09027180
CAS Number: 90890-88-3

Order Now

Specification Test Results

Boiling point 60°C/0.3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR