1-Cyclopropyl-4-piperidinone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Cyclopropylpiperidin-4-one

047918-1G 047918-5G 047918-25G
101881-992EA 76.92 USD
101881-992 101881-994 101881-996
1-Cyclopropyl-4-piperidinone ≥95%
1-Cyclopropyl-4-piperidinone

Hazard: Irritant

Formula: C₈H₁₃NO
MW: 139.20 g/mol
Boiling Pt: 55 °C (2 mmHg)
MDL Number: MFCD07374375
CAS Number: 62813-01-8

Order Now

Specification Test Results

Boiling point 55°C/2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR