1-Hexyltetrahydro-4(1H)-pyridinone ≥95%

Supplier: Matrix Scientific

047915-500MG 047915-1G 047915-5G
101881-980EA 278.46 USD
101881-980 101881-982 101881-984
1-Hexyltetrahydro-4(1H)-pyridinone ≥95%
1-Hexyltetrahydro-4(1H)-pyridinone

Hazard: Irritant

Formula: C₁₁H₂₁NO
Boiling Pt: 100 °C (1 mmHg)
MDL Number: MFCD09027179
CAS Number: 71072-22-5

Order Now

Specification Test Results

Boiling point 100°C/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR