3,7-Dibutyl-3,7-diazabicyclo[3.3.1]nonane ≥95%

Supplier: Matrix Scientific

047775-500MG 047775-1G 047775-5G
101881-374EA 921.54 USD
101881-374 101881-376 101881-378
3,7-Dibutyl-3,7-diazabicyclo[3.3.1]nonane ≥95%
3,7-Dibutyl-3,7-diazabicyclo[3.3.1]nonane

Hazard: Irritant

Formula: C₁₅H₃₀N₂
Boiling Pt: 105 °C (0.3 mmHg)
MDL Number: MFCD09027161
CAS Number: 58324-90-6

Order Now

Specification Test Results

Boiling point 105°C/0.3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR