1-Butyl-4-piperidone ≥95%

Supplier: Matrix Scientific

047771-500MG 047771-1G 047771-5G
101881-350EA 278.46 USD
101881-350 101881-352 101881-354
1-Butyl-4-piperidone ≥95%
1-Butyl-4-piperidone

Hazard: Irritant

Formula: C₉H₁₇NO
Boiling Pt: 50°C/0.1mm
MDL Number: MFCD00023404
CAS Number: 23081-86-9

Order Now

Specification Test Results

Boiling point 50°C/0.1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR