3,7-Diisopropyl-3,7-diazabicyclo[3.3.1]nonan-9-one ≥95%
Supplier: Matrix Scientific
047770-500MG
047770-1G
047770-5G
101881-344EA
278.46
USD
101881-344
101881-346
101881-348
3,7-Diisopropyl-3,7-diazabicyclo[3.3.1]nonan-9-one ≥95%
3,7-Diisopropyl-3,7-diazabicyclo[3.3.1]nonan-9-one
Hazard: Irritant
Formula:
C₁₃H₂₄N₂O Boiling Pt: 112 °C (0.2 mmHg) |
MDL Number:
MFCD09027159 CAS Number: 173973-33-6 |
Specification Test Results
Boiling point | 112°C/0.2mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...