5,6-Dihydro-4H-cyclopenta[b]thiophen-2-ylmethanol ≥95%
Supplier: Matrix Scientific
046418-500MG
046418-1G
101875-414EA
921.54
USD
101875-414
101875-416
5,6-Dihydro-4H-cyclopenta[b]thiophen-2-ylmethanol ≥95%
5,6-Dihydro-4H-cyclopenta[b]thiophen-2-ylmethanol
Hazard: Irritant
Formula:
C₈H₁₀OS MW: 154.23 g/mol Boiling Pt: 48…54 °C (1 mmHg) |
MDL Number:
MFCD08059488 CAS Number: 41301-23-9 |
Specification Test Results
Boiling point | 48-54°C/1mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...