2-[(Dimethylamino)methyl]-6-methoxy-4-methylbenzenol ≥95%
Supplier: Matrix Scientific
046363-500MG
046363-1G
046363-5G
101875-172EA
278.46
USD
101875-172
101875-174
101875-176
2-[(Dimethylamino)methyl]-6-methoxy-4-methylbenzenol ≥95%
2-[(Dimethylamino)methyl]-6-methoxy-4-methylbenzenol
Hazard: Irritant
Formula:
C₁₁H₁₇NO₂ Boiling Pt: 106 °C (0.05 mmHg) |
MDL Number:
MFCD08443958 CAS Number: 123752-64-7 |
Specification Test Results
Boiling point | 106°C/0.05m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...