Methyl-2-(3-amino-4-methoxyphenyl)acetate ≥95%

Supplier: Matrix Scientific

046117-500MG 046117-1G
101874-244EA 461.54 USD
101874-244 101874-246
Methyl-2-(3-amino-4-methoxyphenyl)acetate ≥95%
Methyl-2-(3-amino-4-methoxyphenyl)acetate

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
Boiling Pt: 140 °C (0.3 mmHg)
MDL Number: MFCD12025417
CAS Number: 63304-82-5

Order Now

Specification Test Results

Boiling point 140°C/0.3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR