2-[4-(Trifluoromethoxy)phenoxy]acetonitrile ≥95%

Supplier: Matrix Scientific

046073-500MG 046073-1G 046073-5G
101874-028EA 278.46 USD
101874-028 101874-030 101874-032
2-[4-(Trifluoromethoxy)phenoxy]acetonitrile ≥95%
2-[4-(Trifluoromethoxy)phenoxy]acetonitrile

Hazard: Irritant

Formula: C₉H₆F₃NO₂
Boiling Pt: 119 °C (12 mmHg)
MDL Number: MFCD09152763
CAS Number: 952182-87-5

Order Now

Specification Test Results

Boiling point 119°C/12mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR