4-Nitrostyrene ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Nitro-4-vinylbenzene, 1-Ethenyl-4-nitrobenzene

045975-500MG 045975-1G 045975-5G
101873-548EA 375.38 USD
101873-548 101873-550 101873-552
4-Nitrostyrene ≥95%
4-Nitrostyrene

Hazard: Irritant

Formula: C₈H₇NO₂
MW: 149.15 g/mol
Boiling Pt: 120°C/10mm
MDL Number: MFCD00041254
CAS Number: 100-13-0

Order Now

Specification Test Results

Boiling point 120°C/10mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR