1-Methyl-4-(3-nitro-2-pyridinyl)piperazine ≥95%

Supplier: Matrix Scientific

045919-500MG 045919-1G 045919-5G
101873-264EA 278.46 USD
101873-264 101873-266 101873-268
1-Methyl-4-(3-nitro-2-pyridinyl)piperazine ≥95%
1-Methyl-4-(3-nitro-2-pyridinyl)piperazine

Hazard: Irritant

Formula: C₁₀H₁₄N₄O₂
Boiling Pt: 130 °C (1 mmHg)
MDL Number: MFCD03001301
CAS Number: 5028-15-9

Order Now

Specification Test Results

Boiling point 130°C/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR