3-Nitro-2-(1-pyrrolidinyl)pyridine ≥95%

Supplier: Matrix Scientific

045918-500MG 045918-1G 045918-5G
101873-258EA 278.46 USD
101873-258 101873-260 101873-262
3-Nitro-2-(1-pyrrolidinyl)pyridine ≥95%
3-Nitro-2-(1-pyrrolidinyl)pyridine

Hazard: Irritant

Formula: C₉H₁₁N₃O₂
MW: 193.21 g/mol
Boiling Pt: 102 °C (0.3 mmHg)
MDL Number: MFCD03001300
CAS Number: 26820-73-5

Order Now

Specification Test Results

Boiling point 102°C/0.3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR