1-Boc-isonipecotic acid ethyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl N-BOC-4-piperidinecarboxylate, Ethyl N-Boc-piperidine-4-carboxylate
045889-5G
045889-25G
101873-120EA
15.38
USD
101873-120
101873-122
1-Boc-isonipecotic acid ethyl ester ≥95%
1-Boc-isonipecotic acid ethyl ester
Hazard: Irritant
Formula:
C₁₃H₂₃NO₄ Boiling Pt: 96…98 °C (0.05 mmHg) |
MDL Number:
MFCD01763998 CAS Number: 142851-03-4 |
Specification Test Results
Boiling point | 96-98°C/0.0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...