1-(3-Furyl)-1-ethanol ≥95%

Supplier: Matrix Scientific

045883-500MG 045883-1G
101873-094EA 369.23 USD
101873-094 101873-096
1-(3-Furyl)-1-ethanol ≥95%
1-(3-Furyl)-1-ethanol

Hazard: Irritant

Formula: C₆H₈O₂
Boiling Pt: 42…43 °C (0.7 mmHg)
MDL Number: MFCD02082436
CAS Number: 13129-26-5

Order Now

Specification Test Results

Boiling point 42-43°C/0.7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR