3-[3-(Trifluoromethyl)phenoxy]-2-butanone ≥95%

Supplier: Matrix Scientific

045805-500MG 045805-1G 045805-5G
101872-694EA 278.46 USD
101872-694 101872-696 101872-698
3-[3-(Trifluoromethyl)phenoxy]-2-butanone ≥95%
3-[3-(Trifluoromethyl)phenoxy]-2-butanone

Hazard: Irritant

Formula: C₁₁H₁₁F₃O₂
Boiling Pt: 68 °C (0.2 mmHg)
MDL Number: MFCD00243674
CAS Number: 338975-95-4

Order Now

Specification Test Results

Boiling point 68°C/0.2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR