3-Phenoxy-2-butanone ≥95%

Supplier: Matrix Scientific

045804-500MG 045804-1G 045804-5G
101872-688EA 300 USD
101872-688 101872-690 101872-692
3-Phenoxy-2-butanone ≥95%
3-Phenoxy-2-butanone

Hazard: Irritant

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
Boiling Pt: 46 °C (0.1 mmHg)
MDL Number: MFCD00243672
CAS Number: 6437-85-0

Order Now

Specification Test Results

Boiling point 46°C/0.1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR