N-[(2-Chloro-1,3-thiazol-5-yl)methyl]-N-(cyclopropylmethyl)-1-propanamine ≥95%
Supplier: Matrix Scientific
045784-500MG
045784-1G
045784-5G
101872-578EA
278.46
USD
101872-578
101872-580
101872-582
N-[(2-Chloro-1,3-thiazol-5-yl)methyl]-N-(cyclopropylmethyl)-1-propanamine ≥95%
N-[(2-Chloro-1,3-thiazol-5-yl)methyl]-N-(cyclopropylmethyl)-1-propanamine
Hazard: Irritant
Formula:
C₁₁H₁₇ClN₂S Boiling Pt: 126 °C (2 mmHg) |
MDL Number:
MFCD06659602 CAS Number: 885950-05-0 |
Specification Test Results
Boiling point | 126°C/2mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...