1-[(2-Chloro-1,3-thiazol-5-yl)methyl]azepane ≥95%

Supplier: Matrix Scientific

045783-500MG 045783-1G 045783-5G
101872-572EA 278.46 USD
101872-572 101872-574 101872-576
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]azepane ≥95%
1-[(2-Chloro-1,3-thiazol-5-yl)methyl]azepane

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂S
Boiling Pt: 110 °C (0.5 mmHg)
MDL Number: MFCD06659601
CAS Number: 885950-03-8

Order Now

Specification Test Results

Boiling point 110°C/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR