5-Methylimidazo[1,2-a]pyridine ≥95%

Supplier: Matrix Scientific

045762-500MG 045762-1G 045762-5G
101872-460EA 76.92 USD
101872-460 101872-462 101872-464
5-Methylimidazo[1,2-a]pyridine ≥95%
5-Methylimidazo[1,2-a]pyridine

Hazard: Irritant

Formula: C₈H₈N₂
MW: 132.1600 g/mol
Boiling Pt: 96°C/0.7mm
MDL Number: MFCD06200991
CAS Number: 933-69-7

Order Now

Specification Test Results

Boiling point 96°C/0.7mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR