Methyl-1-[(2-chloro-1,3-thiazol-5-yl)methyl]-4-piperidinecarboxylate ≥95%

Supplier: Matrix Scientific

045665-500MG 045665-1G 045665-5G
101871-998EA 375.38 USD
101871-998 101872-000 101872-002
Methyl-1-[(2-chloro-1,3-thiazol-5-yl)methyl]-4-piperidinecarboxylate ≥95%
Methyl-1-[(2-chloro-1,3-thiazol-5-yl)methyl]-4-piperidinecarboxylate

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₂S
Boiling Pt: 132-134°C/0.01mm
MDL Number: MFCD03001294
CAS Number: 454473-62-2

Order Now

Specification Test Results

Boiling point 132-134°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR