2-Chloro-2,2-difluoro-1-(4-hydroxypiperidino)-1-ethanone ≥95%

Supplier: Matrix Scientific

045652-500MG 045652-1G 045652-5G
101871-936EA 181.54 USD
101871-936 101871-938 101871-940
2-Chloro-2,2-difluoro-1-(4-hydroxypiperidino)-1-ethanone ≥95%
2-Chloro-2,2-difluoro-1-(4-hydroxypiperidino)-1-ethanone

Hazard: Irritant

Formula: C₇H₁₀ClF₂NO₂
Boiling Pt: 117 °C (1 mmHg)
MDL Number: MFCD02082432
CAS Number: 478258-79-6

Order Now

Specification Test Results

Boiling point 117°C/1mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR