Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate ≥95%

Supplier: Matrix Scientific

044395-1G 044395-5G 044395-25G
101865-614EA 56.92 USD
101865-614 101865-616 102833-128
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate ≥95%
Ethyl-2-((dimethylamino)methylene)-3-oxobutanoate

Hazard: Irritant

Formula: C₉H₁₅NO₃
Boiling Pt: 128 °C (0.2 mmHg)
MDL Number: MFCD00087563
CAS Number: 51145-57-4

Order Now

Specification Test Results

Boiling point 128°C/0.2mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR