3-(3-Phenyl-2-propenyl)-2,4-pentanedione ≥95%

Supplier: Matrix Scientific

044241-500MG 044241-1G 044241-5G
101864-890EA 278.46 USD
101864-890 101864-892 101864-894
3-(3-Phenyl-2-propenyl)-2,4-pentanedione ≥95%
3-(3-Phenyl-2-propenyl)-2,4-pentanedione

Hazard: Irritant

Formula: C₁₄H₁₆O₂
MW: 216.28 g/mol
Boiling Pt: 160…164 °C (3 mmHg)
MDL Number: MFCD03425758
CAS Number: 29638-71-9

Order Now

Specification Test Results

Boiling point 160-164°C/3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR