2-(1H-Pyrrol-1-yl)-1-ethanamine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1H-Pyrrol-1-yl)ethanamine

044209-500MG 044209-1G 044209-5G
101864-732EA 269.23 USD
101864-732 101864-734 101864-736
2-(1H-Pyrrol-1-yl)-1-ethanamine ≥95%
2-(1H-Pyrrol-1-yl)-1-ethanamine

Hazard: Irritant

Formula: C₆H₁₀N₂
MW: 110.16 g/mol
Boiling Pt: 60 °C (0.3 mmHg)
MDL Number: MFCD00958089
CAS Number: 29709-35-1

Order Now

Specification Test Results

Boiling point 60°C/0.3mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR