2-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl](methyl)amino]-1-ethanol ≥95%

Supplier: Matrix Scientific

044195-500MG 044195-1G 044195-5G
101864-654EA 387.69 USD
101864-654 101864-656 101864-658
2-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl](methyl)amino]-1-ethanol ≥95%
2-[[3-Chloro-5-(trifluoromethyl)-2-pyridinyl](methyl)amino]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₀ClF₃N₂O
Boiling Pt: 90…92 °C (0.05 mmHg)
MDL Number: MFCD01765376
CAS Number: 263387-09-3

Order Now

Specification Test Results

Boiling point 90-92°C/0.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR