2-[(4-Chlorobenzyl)oxy]-1-ethanol ≥95%

Supplier: Matrix Scientific

044094-500MG 044094-1G 044094-5G
101864-102EA 181.54 USD
101864-102 101864-104 101864-106
2-[(4-Chlorobenzyl)oxy]-1-ethanol ≥95%
2-[(4-Chlorobenzyl)oxy]-1-ethanol

Hazard: Irritant

Formula: C₉H₁₁ClO₂
MW: 186.64 g/mol
Boiling Pt: 108-112°C/0.3mm
MDL Number: MFCD00172032
CAS Number: 1200-15-3

Order Now

Specification Test Results

Boiling point 108-112°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR