2-[3-(Trifluoromethyl)phenoxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

043876-500MG 043876-1G 043876-5G
101863-016EA 556.92 USD
101863-016 101863-018 101863-020
2-[3-(Trifluoromethyl)phenoxy]benzenecarbaldehyde ≥95%
2-[3-(Trifluoromethyl)phenoxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₄H₉F₃O₂
Boiling Pt: 108…110 °C (0.1 mmHg)
MDL Number: MFCD01814776
CAS Number: 408369-24-4

Order Now

Specification Test Results

Boiling point 108-110°C/0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR