Pentafluorophenyl 1-methanesulfonate ≥95%

Supplier: Matrix Scientific

043727-500MG 043727-1G 043727-5G
101862-264EA 375.38 USD
101862-264 101862-266 101862-268
Pentafluorophenyl 1-methanesulfonate ≥95%
Pentafluorophenyl 1-methanesulphonate

Hazard: Irritant

Formula: C₇H₃O₃F₅S
Boiling Pt: 76 °C (0.7 mmHg)
MDL Number: MFCD06659544
CAS Number: 161912-36-3

Order Now

Specification Test Results

Boiling point 76°C/0.7mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR