4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-butyn-2-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-2-(3-hydroxybut-1-yn-1-yl)-5-(trifluoromethyl)pyridine

043641-500MG 043641-1G
101861-830EA 921.54 USD
101861-830 101861-832
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-butyn-2-ol ≥95%
4-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-3-butyn-2-ol

Hazard: Irritant

Formula: C₁₀H₇ClF₃NO
MW: 249.62 g/mol
Boiling Pt: 155°C/0.5mm
MDL Number: MFCD08443949
CAS Number: 937601-45-1

Order Now

Specification Test Results

Boiling point 155°C/0.5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR