2-(Chloromethyl)benzothiazole ≥95%
Supplier: Matrix Scientific
Synonyms:
2-(Chloromethyl)-1,3-benzothiazole
043624-500MG
043624-1G
043624-5G
101861-748EA
33.85
USD
101861-748
101861-750
101861-752
2-(Chloromethyl)benzothiazole ≥95%
2-(Chloromethyl)benzothiazole
Hazard: Irritant/Refrigerate
Formula:
C₈H₆CINS Boiling Pt: 102°C/0.5mm |
MDL Number:
MFCD05664944 CAS Number: 37859-43-1 |
Specification Test Results
Boiling point | 102°C/0.5mm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...