4-Fluorobenzylideneacetone ≥95%
Supplier: Matrix Scientific
Synonyms:
1-(4-Fluorophenyl)but-1-en-3-one
043413-500MG
043413-1G
043413-5G
101860-652EA
204.62
USD
101860-652
101860-654
101860-656
4-Fluorobenzylideneacetone ≥95%
4-Fluorobenzylideneacetone
Hazard: Irritant
Formula:
C₁₀H₉FO Boiling Pt: 119°C/2.5mm |
MDL Number:
MFCD00205144 CAS Number: 1611-38-7 |
Specification Test Results
Boiling point | 119°C/2.5mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...