2-(Chloromethyl)benzoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Chloromethyl)benzo[d]oxazole, 2-(Chloromethyl)-1,3-benzoxazole

043377-1G 043377-5G
101860-484EA 115.38 USD
101860-484 101860-486
2-(Chloromethyl)benzoxazole ≥95%
2-(Chloromethyl)benzoxazole

Hazard: Irritant

Formula: C₈H₆ClNO
MW: 167.5900 g/mol
Boiling Pt: 75°C/0.2mm
MDL Number: MFCD05664964
CAS Number: 41014-43-1

Order Now

Specification Test Results

Boiling point 75°C/0.2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR